Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: foxo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275628.1 | NP_001262557.1 | 5 - 6 | 1244 | 67 | |
NM_142073.5 | NP_650330.3 | 6 - 7 | 1502 | 67 | ||
NM_206482.3 | NP_996204.1 | 6 - 7 | 1369 | 67 | ||
NM_206483.3 | NP_996205.1 | 6 - 7 | 1760 | 67 | ||
GenBank mRNA | AF426831.1 | - | 6 - 7 | 1352 | 67 | |
AY089542.1 | - | 6 - 7 | 1737 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |