Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cv-c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104316.2 | NP_001097786.1 | 7 - 8 | 5242 | 65 | |
NM_001104318.2 | NP_001097788.1 | 8 - 9 | 4575 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |