Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wrd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144598.3 | NP_001138070.1 | 5 - 6 | 1248 | 131 | |
NM_001144599.2 | NP_001138071.1 | 5 - 6 | 1248 | 131 | ||
NM_001144600.2 | NP_001138072.1 | 3 - 4 | 631 | 131 | ||
NM_001260247.1 | NP_001247176.1 | 5 - 6 | 1380 | 131 | ||
NM_001260248.1 | NP_001247177.1 | 5 - 6 | 1207 | 131 | ||
NM_001260249.1 | NP_001247178.1 | 5 - 6 | 1654 | 131 | ||
NM_001260250.1 | NP_001247179.1 | 4 - 5 | 2353 | 131 | ||
NM_001275765.1 | NP_001262694.1 | 6 - 7 | 1797 | 131 | ||
NM_001275766.1 | NP_001262695.1 | 6 - 7 | 1797 | 131 | ||
NM_142424.3 | NP_650681.2 | 5 - 6 | 1421 | 131 | ||
NM_169787.2 | NP_732293.1 | 5 - 6 | 1248 | 131 | ||
NM_169788.1 | NP_732294.1 | 3 - 4 | 750 | 131 | ||
NM_169789.2 | NP_732295.1 | 5 - 6 | 2196 | 131 | ||
NM_169790.1 | NP_732296.1 | 5 - 6 | 2196 | 131 | ||
GenBank mRNA | AY118546.1 | - | 5 - 6 | 2196 | 131 | |
BT021232.1 | - | 5 - 6 | 1247 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |