Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Muc91C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300469.1 | NP_001287398.1 | 1 - 2 | 121 | 68 | |
NM_142487.2 | NP_650744.1 | 1 - 2 | 121 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |