Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170260.2 | NP_001163731.1 | 7 - 8 | 1776 | 62 | |
NM_143150.4 | NP_651407.3 | 7 - 8 | 1970 | 62 | ||
NM_170231.3 | NP_733110.2 | 7 - 8 | 1776 | 62 | ||
GenBank mRNA | AY058286.2 | - | 7 - 8 | 1970 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |