Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ball | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_143251.1 | NP_651508.1 | 2 - 3 | 399 | 64 | |
NM_170292.2 | NP_733171.1 | 1 - 2 | 337 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |