Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tau | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276088.1 | NP_001263017.1 | 6 - 7 | 1569 | 109 | |
NM_001300637.1 | NP_001287566.1 | 6 - 7 | 925 | 109 | ||
NM_001300638.1 | NP_001287567.1 | 5 - 6 | 947 | 109 | ||
NM_001300639.1 | NP_001287568.1 | 7 - 8 | 1405 | 109 | ||
NM_001382126.1 | NP_001369032.1 | 7 - 8 | 1513 | 109 | ||
NM_143318.3 | NP_651575.1 | 6 - 7 | 1387 | 109 | ||
NM_170345.3 | NP_733224.2 | 8 - 9 | 2898 | 109 | ||
GenBank mRNA | AY032977.1 | - | 6 - 7 | 1364 | 109 | |
AY102697.1 | - | 6 - 7 | 1383 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |