Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDase | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276180.1 | NP_001263109.1 | 3 - 4 | 495 | 71 | |
NM_143540.2 | NP_651797.1 | 3 - 4 | 798 | 71 | ||
NM_170488.2 | NP_733367.1 | 3 - 4 | 531 | 71 | ||
NM_170489.2 | NP_733368.1 | 3 - 4 | 903 | 71 | ||
NM_170490.2 | NP_733369.1 | 3 - 4 | 899 | 71 | ||
NM_170491.2 | NP_733370.1 | 4 - 5 | 1034 | 71 | ||
GenBank mRNA | BT150409.1 | - | 3 - 4 | 833 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |