Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pygo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_143615.4 | NP_651872.1 | 2 - 3 | 3326 | 68 | |
GenBank mRNA | AY058500.1 | - | 2 - 3 | 3317 | 68 | |
AY075095.1 | - | 2 - 3 | 2875 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |