Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lk6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_143729.3 | NP_651986.2 | 2 - 3 | 1150 | 60 | |
NM_169423.3 | NP_731624.1 | 2 - 3 | 702 | 60 | ||
GenBank mRNA | BT050542.1 | - | 2 - 3 | 475 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |