Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170106.1 | NP_001163577.1 | 4 - 5 | 1062 | 80 | |
NM_001170107.1 | NP_001163578.1 | 4 - 5 | 1106 | 80 | ||
NM_001300335.1 | NP_001287264.1 | 3 - 4 | 909 | 80 | ||
NM_143731.6 | NP_651988.2 | 4 - 5 | 1111 | 80 | ||
NM_169310.2 | NP_731403.2 | 4 - 5 | 1050 | 80 | ||
GenBank mRNA | AY118517.1 | - | 4 - 5 | 1088 | 80 | |
BT001633.1 | - | 4 - 5 | 1050 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |