Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gel | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043192.3 | NP_001036657.2 | 6 - 7 | 2405 | 88 | |
NM_080126.4 | NP_524865.2 | 7 - 8 | 2657 | 88 | ||
NM_164319.4 | NP_730788.1 | 6 - 7 | 2449 | 88 | ||
NM_164321.5 | NP_730790.2 | 8 - 9 | 2851 | 88 | ||
NM_176394.3 | NP_788571.1 | 7 - 8 | 2628 | 88 | ||
NM_206426.4 | NP_996148.2 | 7 - 8 | 2660 | 88 | ||
NM_206427.4 | NP_996149.3 | 8 - 9 | 2718 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |