Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043211.2 | NP_001036676.1 | 11 - 12 | 3946 | 79 | |
NM_169022.2 | NP_730882.2 | 11 - 12 | 3934 | 79 | ||
GenBank mRNA | BT024186.1 | - | 10 - 11 | 2936 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |