Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PpD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275504.2 | NP_001262433.1 | 3 - 4 | 663 | 71 | |
NM_080207.3 | NP_524946.1 | 4 - 5 | 768 | 71 | ||
NM_169307.2 | NP_731398.1 | 4 - 5 | 633 | 71 | ||
GenBank mRNA | AJ271781.1 | - | 3 - 4 | 582 | 71 | |
AY058356.1 | - | 3 - 4 | 626 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |