Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sap47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_169684.3 | NP_732088.1 | 2 - 3 | 1271 | 85 | |
NM_169685.3 | NP_732089.1 | 3 - 4 | 879 | 85 | ||
NM_169686.3 | NP_732090.1 | 3 - 4 | 1207 | 85 | ||
NM_169687.3 | NP_732091.1 | 3 - 4 | 1025 | 85 | ||
NM_169690.3 | NP_732094.1 | 2 - 3 | 1271 | 85 | ||
GenBank mRNA | AY051871.1 | - | 2 - 3 | 1219 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |