Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tinc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275757.1 | NP_001262686.1 | 3 - 4 | 1050 | 58 | |
NM_169780.5 | NP_732278.2 | 4 - 5 | 1137 | 58 | ||
NM_176509.4 | NP_788686.1 | 4 - 5 | 1137 | 58 | ||
NM_176510.5 | NP_788687.3 | 4 - 5 | 1047 | 58 | ||
NM_176511.3 | NP_788688.2 | 4 - 5 | 1095 | 58 | ||
GenBank mRNA | AF509933.1 | - | 1 - 2 | 312 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |