Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TwdlR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_170282.2 | NP_733161.2 | 1 - 2 | 41 | 62 | |
GenBank mRNA | BT122078.2 | - | 1 - 2 | 41 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |