Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lerp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276067.1 | NP_001262996.1 | 6 - 7 | 866 | 82 | |
NM_170288.3 | NP_733167.2 | 6 - 7 | 881 | 82 | ||
NM_170289.3 | NP_733168.2 | 6 - 7 | 902 | 82 | ||
NM_170290.3 | NP_733169.2 | 6 - 7 | 902 | 82 | ||
GenBank mRNA | AY684632.2 | - | 6 - 7 | 881 | 82 | |
BT021416.1 | - | 6 - 7 | 846 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |