Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079802.3 | NP_524526.1 | 3 - 4 | 1282 | 82 | |
NM_170309.2 | NP_733188.1 | 2 - 3 | 985 | 82 | ||
NM_170312.2 | NP_733191.1 | 2 - 3 | 1204 | 82 | ||
NM_170315.2 | NP_733194.1 | 3 - 4 | 1063 | 82 | ||
NM_170316.2 | NP_733195.1 | 3 - 4 | 1123 | 82 | ||
GenBank mRNA | AF237968.1 | - | 3 - 4 | 202 | 82 | |
AF237969.1 | - | 3 - 4 | 421 | 82 | ||
BT044193.2 | - | 2 - 3 | 1186 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |