Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: side | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144645.3 | NP_001138117.1 | 3 - 4 | 1749 | 62 | |
NM_001260405.2 | NP_001247334.1 | 3 - 4 | 1438 | 62 | ||
NM_001260406.2 | NP_001247335.1 | 3 - 4 | 2171 | 62 | ||
NM_079806.4 | NP_524530.2 | 3 - 4 | 2171 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |