Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhc98D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170288.1 | NP_001163759.1 | 21 - 22 | 6918 | 76 | |
NM_001276107.1 | NP_001263036.1 | 15 - 16 | 4600 | 76 | ||
NM_079817.3 | NP_524541.3 | 22 - 23 | 6939 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |