Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: chp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276205.1 | NP_001263134.1 | 11 - 12 | 3436 | 59 | |
NM_001276206.1 | NP_001263135.1 | 11 - 12 | 3448 | 59 | ||
NM_001276207.1 | NP_001263136.1 | 11 - 12 | 3427 | 59 | ||
NM_001276208.1 | NP_001263137.1 | 11 - 12 | 3448 | 59 | ||
NM_079866.3 | NP_524605.1 | 11 - 12 | 3457 | 59 | ||
GenBank mRNA | BT126010.1 | - | 11 - 12 | 3235 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |