Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bon | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275836.1 | NP_001262765.1 | 4 - 5 | 2787 | 63 | |
NM_001275838.1 | NP_001262767.1 | 4 - 5 | 2787 | 63 | ||
NM_001275839.1 | NP_001262768.1 | 4 - 5 | 2781 | 63 | ||
NM_079985.4 | NP_524724.2 | 4 - 5 | 2787 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |