Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: woc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104476.2 | NP_001097946.1 | 8 - 9 | 4925 | 57 | |
NM_001260397.1 | NP_001247326.1 | 4 - 5 | 1719 | 57 | ||
NM_001276080.1 | NP_001263009.1 | 7 - 8 | 4919 | 57 | ||
NM_001276081.1 | NP_001263010.1 | 8 - 9 | 4922 | 57 | ||
NM_080147.3 | NP_524886.2 | 7 - 8 | 4922 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |