Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acph-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080178.5 | NP_524917.1 | 4 - 5 | 1134 | 73 | |
NM_170453.2 | NP_733332.1 | 3 - 4 | 1079 | 73 | ||
GenBank mRNA | AY071077.1 | - | 4 - 5 | 1131 | 73 | |
BT001641.1 | - | 3 - 4 | 1079 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |