Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275545.1 | NP_001262474.1 | 1 - 2 | 765 | 68 | |
NM_080512.5 | NP_536773.1 | 1 - 2 | 765 | 68 | ||
GenBank mRNA | BT044080.1 | - | 1 - 2 | 761 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |