Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mask | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260351.1 | NP_001247280.1 | 13 - 14 | 6148 | 61 | |
NM_001275974.1 | NP_001262903.1 | 12 - 13 | 4954 | 61 | ||
NM_001275975.1 | NP_001262904.1 | 12 - 13 | 6118 | 61 | ||
NM_001300571.1 | NP_001287500.1 | 11 - 12 | 4915 | 61 | ||
NM_176556.2 | NP_788733.1 | 12 - 13 | 6121 | 61 | ||
NM_176557.2 | NP_788734.1 | 12 - 13 | 6121 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |