Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AhcyL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_206499.2 | NP_996221.1 | 2 - 3 | 393 | 76 | |
NM_206500.2 | NP_996222.1 | 2 - 3 | 270 | 76 | ||
GenBank mRNA | AY113501.1 | - | 2 - 3 | 267 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster |