Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LpR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104463.3 | NP_001097933.2 | 3 - 4 | 755 | 73 | |
NM_001104464.2 | NP_001097934.2 | 3 - 4 | 755 | 73 | ||
NM_001170263.2 | NP_001163734.1 | 3 - 4 | 755 | 73 | ||
NM_001276043.1 | NP_001262972.1 | 3 - 4 | 755 | 73 | ||
NM_001276044.1 | NP_001262973.1 | 4 - 5 | 2975 | 73 | ||
NM_001276045.1 | NP_001262974.1 | 2 - 3 | 249 | 73 | ||
NM_206573.2 | NP_996296.2 | 3 - 4 | 755 | 73 | ||
GenBank mRNA | BT011057.1 | - | 3 - 4 | 755 | 73 | |
BT011063.1 | - | 3 - 4 | 734 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |