Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SkpC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134514.2 | NP_608358.1 | 1 | 492 | 91 | |
GenBank mRNA | BT120007.1 | - | 1 | 426 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |