Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fray | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144605.2 | NP_001138077.1 | 2 | 2328 | 57 | |
NM_001144606.1 | NP_001138078.1 | 2 | 1988 | 57 | ||
NM_001144607.2 | NP_001138079.1 | 2 | 2081 | 57 | ||
NM_001300466.1 | NP_001287395.1 | 4 | 2281 | 57 | ||
NM_001316520.1 | NP_001303449.2 | 2 | 2182 | 57 | ||
NM_001316521.1 | NP_001303450.1 | 3 | 2091 | 57 | ||
NM_057960.4 | NP_477308.1 | 3 | 2346 | 57 | ||
NM_169823.2 | NP_732352.1 | 2 | 2007 | 57 | ||
GenBank mRNA | BT003198.1 | - | 3 | 2342 | 57 | |
BT021945.1 | - | 2 | 2308 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |