Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SkpA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038729.3 | NP_001033818.1 | 1 | 1252 | 72 | |
NM_001297826.1 | NP_001284755.1 | 1 | 534 | 72 | ||
NM_058042.5 | NP_477390.1 | 2 | 795 | 72 | ||
NM_166856.3 | NP_726690.1 | 2 | 643 | 72 | ||
NM_166857.3 | NP_726691.1 | 2 | 758 | 72 | ||
NM_166858.3 | NP_726692.1 | 2 | 1180 | 72 | ||
NM_166859.3 | NP_726693.1 | 2 | 577 | 72 | ||
NM_166860.3 | NP_726694.1 | 2 | 621 | 72 | ||
NM_166861.3 | NP_726695.1 | 2 | 553 | 72 | ||
GenBank mRNA | AY069297.1 | - | 1 | 1214 | 72 | |
BT099582.1 | - | 2 | 581 | 72 | ||
BT099610.1 | - | 2 | 738 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |