Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: edl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079062.3 | NP_523786.2 | 2 | 1275 | 146 | |
GenBank mRNA | AF358670.1 | - | 2 | 1239 | 146 | |
AY069443.1 | - | 2 | 1239 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |