Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kni | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300201.1 | NP_001287130.1 | 3 | 1764 | 142 | |
NM_079463.3 | NP_524187.1 | 3 | 1953 | 142 | ||
GenBank mRNA | AY118798.1 | - | 3 | 1938 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |