Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300614.1 | NP_001287543.1 | 2 | 2363 | 100 | |
NM_079792.3 | NP_524516.2 | 2 | 1604 | 100 | ||
GenBank mRNA | AY051581.1 | - | 2 | 1815 | 100 | |
BT003640.1 | - | 4 | 2571 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |