Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpb8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_141095.3 | NP_649352.1 | 2 | 630 | 91 | |
GenBank mRNA | AY060855.1 | - | 2 | 610 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |