Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lcp65Ag3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144405.2 | NP_652662.1 | 2 | 419 | 104 | |
GenBank mRNA | BT030765.1 | - | 2 | 416 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |