Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsp60C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_135104.5 | NP_608948.2 | 3 | 868 | 77 | |
NM_164654.2 | NP_723104.2 | 3 | 937 | 77 | ||
NM_164655.3 | NP_723105.2 | 2 | 804 | 77 | ||
GenBank mRNA | BT001308.1 | - | 3 | 814 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |