Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RpII15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260170.2 | NP_001247099.1 | 2 | 538 | 133 | |
NM_001300397.1 | NP_001287326.1 | 2 | 545 | 133 | ||
NM_169575.3 | NP_731898.1 | 2 | 545 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |