Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gfat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015157.4 | NP_001015157.2 | 8 - 9 | 1862 | 67 | |
NM_001015158.4 | NP_001015158.1 | 7 - 8 | 1799 | 67 | ||
NM_001015159.4 | NP_001015159.2 | 4 - 5 | 3162 | 67 | ||
NM_001015160.4 | NP_001015160.2 | 8 - 9 | 1826 | 67 | ||
NM_001015161.4 | NP_001015161.2 | 6 - 7 | 1775 | 67 | ||
NM_001015162.4 | NP_001015162.1 | 8 - 9 | 1726 | 67 | ||
NM_001110995.2 | NP_001104465.1 | 7 - 8 | 2037 | 67 | ||
NM_001201439.2 | NP_001188368.1 | 9 - 10 | 1974 | 67 | ||
NM_001201440.1 | NP_001188369.1 | 8 - 9 | 2064 | 67 | ||
GenBank mRNA | BT082094.1 | - | 4 - 5 | 3123 | 67 | |
BT120108.1 | - | 8 - 9 | 1683 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |