Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spase22-23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260350.2 | NP_001247279.1 | 1 | 1446 | 132 | |
GenBank mRNA | AY070615.1 | - | 1 | 1443 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |