Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dbp80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015151.3 | NP_001015151.1 | 1 - 2 | 103 | 102 | |
NM_001015152.4 | NP_001015152.2 | 1 - 2 | 103 | 102 | ||
NM_001276239.1 | NP_001263168.1 | 1 - 2 | 103 | 102 | ||
NM_001276240.1 | NP_001263169.1 | 1 - 2 | 103 | 102 | ||
NM_001276241.1 | NP_001263170.1 | 1 - 2 | 103 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |