Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UQCR-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110937.3 | NP_001104407.2 | 2 - 3 | 172 | 110 | |
NM_001170381.2 | NP_001163852.1 | 2 - 3 | 156 | 110 | ||
NM_001170382.1 | NP_001163853.1 | 2 - 3 | 174 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |