Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Haspin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015349.4 | NP_001015349.2 | 3 - 4 | 1246 | 89 | |
GenBank mRNA | AY118897.1 | - | 3 - 4 | 1274 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |