Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cep43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002125.1 | NP_001002125.1 | 7 - 8 | 535 | 66 | |
XM_005160662.4 | XP_005160719.1 | 7 - 8 | 569 | 66 | ||
XM_005160663.4 | XP_005160720.1 | 7 - 8 | 569 | 66 | ||
XM_005160664.4 | XP_005160721.1 | 7 - 8 | 569 | 66 | ||
XM_017352461.2 | XP_017207950.1 | 7 - 8 | 569 | 66 | ||
XM_021468390.1 | XP_021324065.1 | 7 - 8 | 569 | 66 | ||
XM_021468391.1 | XP_021324066.1 | 7 - 8 | 569 | 66 | ||
XM_021468392.1 | XP_021324067.1 | 7 - 8 | 569 | 66 | ||
XM_021468393.1 | XP_021324068.1 | 7 - 8 | 569 | 66 | ||
GenBank mRNA | BC071450.1 | - | 7 - 8 | 535 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |