Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cenpp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003555.3 | NP_001003555.2 | 5 - 6 | 555 | 128 | |
GenBank mRNA | BC078335.1 | - | 5 - 6 | 529 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |