Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prc1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004494.1 | NP_001004494.1 | 14 - 15 | 1825 | 73 | |
GenBank mRNA | BC075789.1 | - | 14 - 15 | 1825 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |