Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zpcx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012244.2 | NP_001012244.2 | 6 - 7 | 1739 | 77 | |
GenBank mRNA | AY737480.1 | - | 6 - 7 | 1679 | 77 | |
BC109471.1 | - | 6 - 7 | 1739 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |