Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nsfb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024454.2 | NP_001019625.2 | 6 - 7 | 565 | 66 | |
NM_001354163.1 | NP_001341092.1 | 6 - 7 | 565 | 66 | ||
XM_005156207.4 | XP_005156264.1 | 6 - 7 | 602 | 66 | ||
XM_017358461.2 | XP_017213950.1 | 5 - 6 | 412 | 66 | ||
GenBank mRNA | BC162793.1 | - | 6 - 7 | 543 | 66 | |
BC162811.1 | - | 6 - 7 | 543 | 66 | ||
DQ022212.1 | - | 6 - 7 | 565 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |