Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ripply1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039108.1 | NP_001034197.1 | 1 - 2 | 155 | 83 | |
GenBank mRNA | AB212219.1 | - | 1 - 2 | 155 | 83 | |
BC162410.1 | - | 1 - 2 | 112 | 83 | ||
BC162427.1 | - | 1 - 2 | 112 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |